Learn More
Med Vet International Pinnacle I.N. (intranasal) Streptococcus Equi Vaccine, LIVe Culture, 10 X 2.5mL
Catalog No. 502828002
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Pinnacle I.N. Streptococcus Equi Vaccine, Live Culture, for intranasal use only. For the vaccination of healthy horses 9 months of age or older as an aid in the prevention of disease (strangles) caused by Streptococcus equi. Meat withdrawal: 30 days.
Catalog No. 50-282-8002 Supplier Med Vet International Supplier No. RXPINNACLEIN
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Pinnacle I.N. Streptococcus Equi Vaccine, Live Culture, for intranasal use only. For the vaccination of healthy horses 9 months of age or older as an aid in the prevention of disease (strangles) caused by Streptococcus equi. Meat withdrawal: 30 days.