Learn More
Medchemexpress LLC Pnd-1186 hydrochloride | 1356154-94-3 | 99.9% | 537.96 | 100 MG
SDP
Catalog No. 5000348206
Encompass_Preferred
PND-1186 hydrochloride (VS-4718 hydrochloride) is a potent, highly-specific, and reversible inhibitor of FAK (Focal Adhesion Kinase) with an IC50 of 1.5 nM. This compound has demonstrated the ability to selectively promote tumor cell apoptosis. It is intended for research use only.
  • Potent, highly-specific, and reversible fak inhibitor.
  • Selectively promotes tumor cell apoptosis.
  • Effective in inhibiting fak Tyr-397 phosphorylation in breast carcinoma cells.
  • Shown to inhibit 4t1 subcutaneous tumor growth in animal models by inducing apoptosis.
Catalog No. 50-003-48206 Supplier Medchemexpress LLC Supplier No. HY13917A100MG
PND-1186 hydrochloride (VS-4718 hydrochloride) is a potent, highly-specific, and reversible inhibitor of FAK (Focal Adhesion Kinase) with an IC50 of 1.5 nM. This compound has demonstrated the ability to selectively promote tumor cell apoptosis. It is intended for research use only.
  • Potent, highly-specific, and reversible fak inhibitor.
  • Selectively promotes tumor cell apoptosis.
  • Effective in inhibiting fak Tyr-397 phosphorylation in breast carcinoma cells.
  • Shown to inhibit 4t1 subcutaneous tumor growth in animal models by inducing apoptosis.