Learn More
Medchemexpress LLC PND-1186 hydrochloride | 1356154-94-3 | 99.9% | 537.96 | 25 MG
SDP
Catalog No. 5000349548
Encompass_Preferred
PND-1186 hydrochloride is a potent, highly-specific, and reversible inhibitor of FAK with an IC50 of 1.5 nM. It selectively promotes tumor cell apoptosis.
  • Potent, highly-specific, and reversible inhibitor of FAK
  • Selectively promotes tumor cell apoptosis
  • Inhibits FAK Tyr-397 phosphorylation
  • Elevates total FAK protein levels
  • Inhibits 4T1 subcutaneous tumor growth
  • Induces apoptosis in BALB/c mice
Catalog No. 50-003-49548 Supplier Medchemexpress LLC Supplier No. 25MGHY13917A
PND-1186 hydrochloride is a potent, highly-specific, and reversible inhibitor of FAK with an IC50 of 1.5 nM. It selectively promotes tumor cell apoptosis.
  • Potent, highly-specific, and reversible inhibitor of FAK
  • Selectively promotes tumor cell apoptosis
  • Inhibits FAK Tyr-397 phosphorylation
  • Elevates total FAK protein levels
  • Inhibits 4T1 subcutaneous tumor growth
  • Induces apoptosis in BALB/c mice