Learn More
Medchemexpress LLC PND-1186 hydrochloride | 1356154-94-3 | 99.9% | 537.96 | 50 MG
SDP
Catalog No. 5000348385
Encompass_Preferred
PND-1186 hydrochloride is a potent, highly-specific, and reversible inhibitor of FAK with an IC50 of 1.5 nM. It selectively promotes tumor cell apoptosis.
  • Potent, highly-specific, and reversible FAK inhibitor.
  • IC50 of 1.5 nM for FAK.
  • Selectively promotes tumor cell apoptosis.
  • Inhibits FAK Tyr-397 phosphorylation in breast carcinoma cells.
  • Reduces tumor growth by inducing apoptosis in vivo.
Catalog No. 50-003-48385 Supplier Medchemexpress LLC Supplier No. HY13917A50MG
PND-1186 hydrochloride is a potent, highly-specific, and reversible inhibitor of FAK with an IC50 of 1.5 nM. It selectively promotes tumor cell apoptosis.
  • Potent, highly-specific, and reversible FAK inhibitor.
  • IC50 of 1.5 nM for FAK.
  • Selectively promotes tumor cell apoptosis.
  • Inhibits FAK Tyr-397 phosphorylation in breast carcinoma cells.
  • Reduces tumor growth by inducing apoptosis in vivo.