Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PND-1186 hydrochloride | 1356154-94-3 | 99.9% | C25H27ClF3N5O3 | 10MG
SDP

Catalog No. 5000260967
Encompass_Preferred

PND-1186 hydrochloride is the hydrochloride salt of a potent, selective, and reversible focal adhesion kinase (FAK) inhibitor supplied for laboratory research. It exhibits nanomolar potency (IC50 = 1.5 nM) and selectively promotes tumor cell apoptosis. Supplied as a high-purity solid with defined solubility and storage recommendations for controlled experimental use.

  • Potent FAK inhibition with IC50 of 1.5 nM.
  • High purity (99.9%).
  • Molecular formula C25H27ClF3N5O3; molecular weight 537.96.
  • Soluble in DMSO (100 mg/mL) and water (20 mg/mL).
  • Recommended storage: 4°C for solid; in solvent -80°C up to 6 months.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-60967 Supplier Medchemexpress LLC Supplier No. HY13917A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PND-1186 hydrochloride is the hydrochloride salt of a potent, selective, and reversible focal adhesion kinase (FAK) inhibitor supplied for laboratory research. It exhibits nanomolar potency (IC50 = 1.5 nM) and selectively promotes tumor cell apoptosis. Supplied as a high-purity solid with defined solubility and storage recommendations for controlled experimental use.

  • Potent FAK inhibition with IC50 of 1.5 nM.
  • High purity (99.9%).
  • Molecular formula C25H27ClF3N5O3; molecular weight 537.96.
  • Soluble in DMSO (100 mg/mL) and water (20 mg/mL).
  • Recommended storage: 4°C for solid; in solvent -80°C up to 6 months.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.