Learn More
Selleck Chemical LLC PND-1186 (VS-4718) 5mg 1061353-68-1 SR-2156
SDP
Supplier:  Selleck Chemical LLC S76535MG
Encompass_Preferred
PND-1186 (VS-4718, SR-2156) is a reversible and selective FAK inhibitor with IC50 of 1.5 nM. PND-1186 selectively promotes tumor cell apoptosis. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0881241