Learn More
ATCC PNEC30; Prostate Neuroendocrine; Mouse (Mus musculus)
Supplier:  ATCC CRL2930
Encompass
PNEC30 is a neuroendocrine cell line that was isolated in 2002 from the prostate of a 6-month-old male mouse. The cell line was established from CR2-TAg prostate tumors and metastases. It can be used for genetic and/or pharmacologic studies of the regulation of NE cell proliferation, differentiation, and tumorigenesis.
Catalog No. 50-238-2563