Learn More
STEMCELL Technologies PneumaCult™-ALI Medium with 6.5 mm Transwell Inserts, Size: 1 Kit
SDP
Catalog No. NC1382950
Encompass
Medium and Transwell inserts for human airway epithelial cells cultured at the air-liquid interface
Catalog No. NC1382950 Supplier STEMCELL Technologies Supplier No. 05022
Medium and Transwell inserts for human airway epithelial cells cultured at the air-liquid interface