Learn More
STEMCELL Technologies PneumaCult™-Ex Medium, Size: 500 mL
SDP
Catalog No. NC0867011
Encompass
Serum- and BPE-free medium for expansion of primary human airway epithelial cells
Catalog No. NC0867011 Supplier STEMCELL Technologies Supplier No. 05008
Serum- and BPE-free medium for expansion of primary human airway epithelial cells