Learn More
Ampco Pneumatic Scaling Square Shank Chisel with 5/8 in. Blade
SDP
Catalog No. 19173585 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
5 in.
Catalog No. Length (English)
19-173-585 5 in.
1 options
Catalog No. 19-173-585 Supplier Ampco Supplier No. CS23ST
Add to Cart
Add to Cart
Pneumatic scaling chisel bit tailored for spark-free scraping and chipping.
  • See image for additional information.
  • Blade width: 5/8 in.
  • Chisel diameter (d): 1/2 in.
  • Length: 5 in.
Specifications
Product Type Pneumatic Scaling Chisel
Length (English) 5 in.
Quantity 1 Ea.