Learn More
Grainger Legris Pneumatic Threshold Sensor, Polymer Body, 3/8 in. NPT, 5/32 in. Tube, 1 ea.
Catalog No. 502784257
Encompass_Preferred
The pneumatic threshold sensor detects the absence of pressure and translates it into a high-pressure pneumatic output. It monitors the decaying or exhausting side of a pneumatic cylinder piston to signal the end of a stroke, enabling the initiation of subsequent system operations. Designed for accuracy, the unit can be mounted on cylinders, control valves, or terminal blocks positioned between the cylinder and flow control regulator.
  • Compact design
  • Suitable for use with compressed air
  • Working pressure range of 43.5 psi to 116 psi
  • Operating temperature range of 5°F to 140°F
  • Replaces traditional sequence valves or limit switches
  • Eliminates ongoing adjustment associated with standard limit valves
Catalog No. 50-278-4257 Supplier Grainger Supplier No. 19H013
The pneumatic threshold sensor detects the absence of pressure and translates it into a high-pressure pneumatic output. It monitors the decaying or exhausting side of a pneumatic cylinder piston to signal the end of a stroke, enabling the initiation of subsequent system operations. Designed for accuracy, the unit can be mounted on cylinders, control valves, or terminal blocks positioned between the cylinder and flow control regulator.
  • Compact design
  • Suitable for use with compressed air
  • Working pressure range of 43.5 psi to 116 psi
  • Operating temperature range of 5°F to 140°F
  • Replaces traditional sequence valves or limit switches
  • Eliminates ongoing adjustment associated with standard limit valves