Learn More
Grainger Cleveland Vibrator Company Turbomite Pneumatic Turbine Vibrator, Acetal, 11.4 lbf, 1 ea
Catalog No. 502785897
Encompass_Preferred
This pneumatic turbine vibrator features a construction of acetal plastic and laser-etched 304 stainless steel, designed for continuous duty applications in chutes and pipes. The unit is engineered for high efficiency, providing optimal force-to-weight and force-to-air consumption ratios while maintaining low noise levels.
  • Permanently lubricated roller bearings
  • Includes grade 5 mounting hardware and muffler
  • Operates at up to 20,000 VPM at 80 psi
  • Maximum force output of 11.4 lbf
  • Designed for materials weighing 25 lb. to 100 lb.
  • Maximum operating temperature of 185°F
Catalog No. 50-278-5897 Supplier Grainger Supplier No. 18K887
This pneumatic turbine vibrator features a construction of acetal plastic and laser-etched 304 stainless steel, designed for continuous duty applications in chutes and pipes. The unit is engineered for high efficiency, providing optimal force-to-weight and force-to-air consumption ratios while maintaining low noise levels.
  • Permanently lubricated roller bearings
  • Includes grade 5 mounting hardware and muffler
  • Operates at up to 20,000 VPM at 80 psi
  • Maximum force output of 11.4 lbf
  • Designed for materials weighing 25 lb. to 100 lb.
  • Maximum operating temperature of 185°F