Learn More
Grainger Cleveland Vibrator Company CVT-P-60 Pneumatic Turbine Vibrator, Acetal Plastic, 147 Lb, 6,560 Vpm, 1 ea
Catalog No. 502784978
Encompass_Preferred
The CVT-P-60 is a quiet pneumatic turbine vibrator engineered for clean room and sterile environments. Constructed from durable acetal plastic with laser-etched #304 stainless steel, this vibrator provides efficient force production for small-scale applications, including bins, hoppers, chutes, and packaging feeders.
  • Force output range from 7 lbf. to 146.8 lbf.
  • Suitable for materials weighing between 25 lbs. and 1,000 lbs.
  • Features permanently lubricated and sealed bearings for continuous duty.
  • Operates at a remarkably low noise level of 76 dBA at 80 psi.
  • Equipped with 1/4 in. NPT air inlet and outlet ports.
  • Maximum operating temperature of 185°F.
  • Includes grade 5 mounting hardware and an integrated muffler.
Catalog No. 50-278-4978 Supplier Grainger Supplier No. 18K890
The CVT-P-60 is a quiet pneumatic turbine vibrator engineered for clean room and sterile environments. Constructed from durable acetal plastic with laser-etched #304 stainless steel, this vibrator provides efficient force production for small-scale applications, including bins, hoppers, chutes, and packaging feeders.
  • Force output range from 7 lbf. to 146.8 lbf.
  • Suitable for materials weighing between 25 lbs. and 1,000 lbs.
  • Features permanently lubricated and sealed bearings for continuous duty.
  • Operates at a remarkably low noise level of 76 dBA at 80 psi.
  • Equipped with 1/4 in. NPT air inlet and outlet ports.
  • Maximum operating temperature of 185°F.
  • Includes grade 5 mounting hardware and an integrated muffler.