Learn More
Grainger Pneumatic Vibrator 160 lb 5500 vpm
Catalog No. 502750258
Encompass_Preferred
The VIBCO Pneumatic Vibrator 160 lbf is a pneumatically powered concrete vibrator that produces 160 pounds of force. It has a 1-1/2 inch diameter head and is powered by compressed air at 80-100 PSI. The vibrator has a flexible shaft that allows it to reach tight spaces and consolidate concrete. It has an air consumption of 18-22 CFM and is designed for concrete consolidation in forms, around rebar, and in block fill applications. The flexible shaft is 4 feet long and constructed of high quality steel. This pneumatic vibrator provides efficient, economical vibration for concrete work.
Catalog No. 50-275-0258 Supplier Grainger Supplier No. 14K262
The VIBCO Pneumatic Vibrator 160 lbf is a pneumatically powered concrete vibrator that produces 160 pounds of force. It has a 1-1/2 inch diameter head and is powered by compressed air at 80-100 PSI. The vibrator has a flexible shaft that allows it to reach tight spaces and consolidate concrete. It has an air consumption of 18-22 CFM and is designed for concrete consolidation in forms, around rebar, and in block fill applications. The flexible shaft is 4 feet long and constructed of high quality steel. This pneumatic vibrator provides efficient, economical vibration for concrete work.