Learn More
Grainger Pneumatic Vibrator 20 lb 12 000 vpm
Catalog No. 502750582
Encompass_Preferred
Grainger pneumatic vibrators enhance material flow and prevent buildup in bins, hoppers, and chutes using compressed air to power an internal piston. Features include 7 models with 133-4448 lbs force, aluminum, steel, and rubber construction, 40-150 PSI air pressure, rotary and ball bearing options, adjustable mounts, vibration isolation, weights from 3.5-39 lbs, and a 1-year warranty. These compact vibrators offer heavy-duty performance for industrial material handling.
Catalog No. 50-275-0582 Supplier Grainger Supplier No. 14K260
Grainger pneumatic vibrators enhance material flow and prevent buildup in bins, hoppers, and chutes using compressed air to power an internal piston. Features include 7 models with 133-4448 lbs force, aluminum, steel, and rubber construction, 40-150 PSI air pressure, rotary and ball bearing options, adjustable mounts, vibration isolation, weights from 3.5-39 lbs, and a 1-year warranty. These compact vibrators offer heavy-duty performance for industrial material handling.