Learn More
Grainger Cleveland Vibrator VM-38 Miniature Piston Vibrator, 304 Stainless Steel, 11,400 VPM, 1 ea.
Catalog No. 502784924
Encompass_Preferred
The VM-38 miniature pneumatic piston vibrator is designed for intermittent duty in applications requiring small force outputs and high frequencies. Featuring a compact, spring-less design and a #304 stainless steel housing, this unit is suitable for use on small bins, hoppers, packaging and filling machines, and parts tracks.
  • Constructed with a dual-diameter piston that allows for mounting at any angle
  • Provides a maximum force of 21 lbf. at 80 psi
  • Achieves a vibration frequency of 11,400 VPM at 80 psi
  • Compatible with optional exhaust muffler to reduce noise levels
  • Includes mounting hardware and push-fit hose connection for efficient installation
  • Suitable for operation in hazardous areas due to pneumatic power source
  • Rated for a maximum temperature of 200°F
Catalog No. 50-278-4924 Supplier Grainger Supplier No. 18K852
The VM-38 miniature pneumatic piston vibrator is designed for intermittent duty in applications requiring small force outputs and high frequencies. Featuring a compact, spring-less design and a #304 stainless steel housing, this unit is suitable for use on small bins, hoppers, packaging and filling machines, and parts tracks.
  • Constructed with a dual-diameter piston that allows for mounting at any angle
  • Provides a maximum force of 21 lbf. at 80 psi
  • Achieves a vibration frequency of 11,400 VPM at 80 psi
  • Compatible with optional exhaust muffler to reduce noise levels
  • Includes mounting hardware and push-fit hose connection for efficient installation
  • Suitable for operation in hazardous areas due to pneumatic power source
  • Rated for a maximum temperature of 200°F