Learn More
Grainger Cleveland Vibrator Co. Turbine Vibrator, Acetal, Universal Mount, 24.8 lbf Max Force, 1 ea
Catalog No. 502785449
Encompass_Preferred
The pneumatic turbine vibrator is a compact, high-efficiency device engineered for precise material movement in small bins, hoppers, chutes, and feeders. Designed for demanding environments, including food and pharmaceutical clean rooms, this unit features a durable acetal plastic and #304 stainless steel construction that ensures long-term reliability. Its turbine design allows for quiet, continuous-duty operation with exceptionally low air consumption, effectively eliminating the need for periodic lubrication.
  • Designed for materials between 25 lb. and 200 lb.
  • Operates at up to 20,900 VPM at 80 psi.
  • Features permanently lubricated roller bearings.
  • Constructed with acetal plastic and laser-etched #304 stainless steel.
  • Universal mounting capabilities for flexible installation.
  • Includes Grade 5 mounting hardware.
  • Maximum operating temperature of 185°F.
Catalog No. 50-278-5449 Supplier Grainger Supplier No. 18K888
The pneumatic turbine vibrator is a compact, high-efficiency device engineered for precise material movement in small bins, hoppers, chutes, and feeders. Designed for demanding environments, including food and pharmaceutical clean rooms, this unit features a durable acetal plastic and #304 stainless steel construction that ensures long-term reliability. Its turbine design allows for quiet, continuous-duty operation with exceptionally low air consumption, effectively eliminating the need for periodic lubrication.
  • Designed for materials between 25 lb. and 200 lb.
  • Operates at up to 20,900 VPM at 80 psi.
  • Features permanently lubricated roller bearings.
  • Constructed with acetal plastic and laser-etched #304 stainless steel.
  • Universal mounting capabilities for flexible installation.
  • Includes Grade 5 mounting hardware.
  • Maximum operating temperature of 185°F.