Learn More
Apexbio Technology LLC Pneumocandin B0 135575-42-7 1mg
SDP
Catalog No. 5000089539
Encompass_Preferred
Pneumocandin B0 (CAS 135575-42-7) is a small-molecule inhibitor targeting -(1 3)-D-glucan synthase It is designed to inhibit this enzyme thereby disrupting fungal cell wall biosynthesis Pneumocandin B0 exerts its biological activity primarily through inhibition of -(1 3)-D-glucan synthase In experimental studies Pneumocandin B0 demonstrates effective inhibition across multiple fungal strains with IC50 values typically observed in the low nanomolar range Based on these pharmacological properties Pneumocandin B0 holds research potential as a reference compound for studying fungal cell wall biosynthesis pathways and as a lead compound in antifungal drug discovery programs
Catalog No. 50-000-89539 Supplier Apexbio Technology LLC Supplier No. B47241
Pneumocandin B0 (CAS 135575-42-7) is a small-molecule inhibitor targeting -(1 3)-D-glucan synthase It is designed to inhibit this enzyme thereby disrupting fungal cell wall biosynthesis Pneumocandin B0 exerts its biological activity primarily through inhibition of -(1 3)-D-glucan synthase In experimental studies Pneumocandin B0 demonstrates effective inhibition across multiple fungal strains with IC50 values typically observed in the low nanomolar range Based on these pharmacological properties Pneumocandin B0 holds research potential as a reference compound for studying fungal cell wall biosynthesis pathways and as a lead compound in antifungal drug discovery programs