Learn More
Diagnostic Biosystems PNEUMOCYSTIS CARINII Pneumocystis carinii
Catalog No. 502407549
Encompass_Preferred
This antibody reacts with P. carinii in the fixed human tissues. The antibody is specific against an 82 kDa parasite specific polypeptide. It does not crossreact with G. lamblia, T. gondii, T. cruzi, L. tropica, E. histolytica, C. albicans and P. falciparum.
Catalog No. 50-240-7549 Supplier Diagnostic Biosystems Supplier No. PDM584
This antibody reacts with P. carinii in the fixed human tissues. The antibody is specific against an 82 kDa parasite specific polypeptide. It does not crossreact with G. lamblia, T. gondii, T. cruzi, L. tropica, E. histolytica, C. albicans and P. falciparum.