Learn More
New England Biolabs, Inc. PNGase A – 750 units
SDP
Catalog No. 502024049
Encompass
PNGase A cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and short complex oligosaccharides such as those found in plant and insect cells from N-linked glycoproteins and glycopeptides. PNGase A differs from PNGase F in that it cleaves N-linked glycans with or without (1,3)-linked core fucose residues.
Catalog No. 50-202-4049 Supplier New England Biolabs, Inc. Supplier No. P0707L
PNGase A cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and short complex oligosaccharides such as those found in plant and insect cells from N-linked glycoproteins and glycopeptides. PNGase A differs from PNGase F in that it cleaves N-linked glycans with or without (1,3)-linked core fucose residues.