Learn More
GENOVIS INC PNGase F Automation 96 x 50 units
Catalog No. NC3666181
Encompass
Lyophilized enzyme in 96-well plates for automated hydrolysis of N-glycans on 96 x 50 ��g glycoprotein
Catalog No. NC3666181 Supplier GENOVIS INC Supplier No. G1PFA050 96 X 50
Lyophilized enzyme in 96-well plates for automated hydrolysis of N-glycans on 96 x 50 ��g glycoprotein