Learn More
New England Biolabs, Inc. PNGase F (Glycerol-free), Recombinant – 15000 units
SDP
Supplier:  New England Biolabs, Inc. P0709S
Encompass
PNGase F is the most effective enzymatic method for removing almost all N-linked oligosaccharides from glycoproteins. PNGase F (Glycerol-free), Recombinant is a recombinant amidase, which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides.
Catalog No. 50-591-170