Learn More
New England Biolabs, Inc. PNGase F – 75000 units
SDP
Catalog No. 50811829
Encompass
PNGase F is the most effective enzymatic method for removing almost all N-linked oligosaccharides from glycoproteins. PNGase F is an amidase, which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides.
  • Leaves N-glycan core oligosaccharides intact and suitable for further analysis
  • Non-recombinant with no detectable endoglycosidase F1, F2 or F3 contamination
  • >= 95% purity, as determined by SDS-PAGE and intact ESI-MS
  • Stored in 50% glycerol
  • Optimal activity and stability for up to 24 months
  • Can be used under native or denaturing conditions
Catalog No. 50-811-829 Supplier New England Biolabs, Inc. Supplier No. P0704L
PNGase F is the most effective enzymatic method for removing almost all N-linked oligosaccharides from glycoproteins. PNGase F is an amidase, which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides.
  • Leaves N-glycan core oligosaccharides intact and suitable for further analysis
  • Non-recombinant with no detectable endoglycosidase F1, F2 or F3 contamination
  • >= 95% purity, as determined by SDS-PAGE and intact ESI-MS
  • Stored in 50% glycerol
  • Optimal activity and stability for up to 24 months
  • Can be used under native or denaturing conditions