Learn More
Agilent Technologies Glyko PNGase F (native), 100 mU
Catalog No. NC9285718
Encompass
Native PNGase F, peptide-N-glycosidase F, peptide-N4-(N-acetyl-β-glucosaminyl)asparagine amidase. Releases intact N-glycans by cleaving between the innermost GlcNAc and Asn. PNGase F releases intact N-linked oligosaccharides from glycoproteins and glycopeptides. The enzyme will not cleave oligosaccharides from a single asparagine residue and cleavage does not occur if there is core α1-3 linked fucose as commonly encountered in plant glycoproteins Phosphate, sulfate and sialic acid groups attached to the oligosaccharide do not affect cleavage.
Catalog No. NC9285718 Supplier Agilent Technologies Supplier No. GKE5003
Native PNGase F, peptide-N-glycosidase F, peptide-N4-(N-acetyl-β-glucosaminyl)asparagine amidase. Releases intact N-glycans by cleaving between the innermost GlcNAc and Asn. PNGase F releases intact N-linked oligosaccharides from glycoproteins and glycopeptides. The enzyme will not cleave oligosaccharides from a single asparagine residue and cleavage does not occur if there is core α1-3 linked fucose as commonly encountered in plant glycoproteins Phosphate, sulfate and sialic acid groups attached to the oligosaccharide do not affect cleavage.