Learn More
Bulldog Bio Inc PNGase F PRIME Glycosidase 500UL
SDP
Supplier:  Bulldog Bio Inc NZPP050
Encompass_Preferred
LD: Manufactured by N-Zyme Scientifics and supplied by Bulldog Bio Inc. Reagent; Molecular Biology; PNGase F PRIME, Glycosidase Specific for N-linked Oligosaccharides (Mannose, Hybrid, and Complex), 500 units/ul, 2x 50ul vials, 50,000 IUB milliunts
Catalog No. 50-999-475