Learn More
Bulldog Bio Inc PNGase F PRIME-LY, 1 x 100ug lyophillized in vial (1,000 units/ µl). 50 µl yields 50,000 units. 
SDP
Catalog No. NC1629843
Encompass_Preferred
Lyophillized PNGase F PRIME is a mutant recombinant PNGase F cloned from Flavobacterium meningosepticum and expressed and purified from E. coli. The proprietary changes made to PNGase F have been shown to have unique characteristics when compared to other commercially-available sources of PNGase F.
Catalog No. NC1629843 Supplier Bulldog Bio Inc Supplier No. NZPP010LY
Lyophillized PNGase F PRIME is a mutant recombinant PNGase F cloned from Flavobacterium meningosepticum and expressed and purified from E. coli. The proprietary changes made to PNGase F have been shown to have unique characteristics when compared to other commercially-available sources of PNGase F.