Learn More
Bulldog Bio Inc PNGase F PRIME-LY, Glycosidase, 2 x 100 μg vials, lyophilized, total activity 100,000 units, purity ≥95%
SDP
Catalog No. NC2228832
Encompass_Preferred
PNGase F PRIME-LY, Glycosidase, 2 x 100 μg vials, lyophilized, total activity 100,000 units, purity ≥95%
This item is not returnable. View return policy
Catalog No. NC2228832 Supplier Bulldog Bio Inc Supplier No. NZPP050LY
PNGase F PRIME-LY, Glycosidase, 2 x 100 μg vials, lyophilized, total activity 100,000 units, purity ≥95%