Learn More
New England Biolabs, Inc. PNGase F, Recombinant – 15000 units
SDP
Catalog No. 50591168
Encompass
PNGase F is the most effective enzymatic method for removing almost all N-linked oligosaccharides from glycoproteins. PNGase F, Recombinant is a recombinant amidase, which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides.
Catalog No. 50-591-168 Supplier New England Biolabs, Inc. Supplier No. P0708S
PNGase F is the most effective enzymatic method for removing almost all N-linked oligosaccharides from glycoproteins. PNGase F, Recombinant is a recombinant amidase, which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides.