Learn More
New England Biolabs, Inc. PNGase F, Recombinant – 75000 units
SDP
Catalog No. 50591167
Encompass
PNGase F is the most effective enzymatic method for removing almost all N-linked oligosaccharides from glycoproteins. PNGase F, Recombinant is a recombinant amidase, which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides.
Catalog No. 50-591-167 Supplier New England Biolabs, Inc. Supplier No. P0708L
PNGase F is the most effective enzymatic method for removing almost all N-linked oligosaccharides from glycoproteins. PNGase F, Recombinant is a recombinant amidase, which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides.