Learn More
Cayman Chemical p-nitro-Cyclic Pifithrin-α, Purity: ≥95%, CAS: 60477-38-5, 10 mg
SDP
Catalog No. 502965661
Encompass_Preferred
A cell-permeable form of cyclic PFT-α; 10-fold more active that PFT-α in protecting cortical neurons exposed to etoposide (ED50 = 30 nM); not active in vivo
Catalog No. 50-296-5661 Supplier Cayman Chemical Supplier No. 1784810 MG
A cell-permeable form of cyclic PFT-α; 10-fold more active that PFT-α in protecting cortical neurons exposed to etoposide (ED50 = 30 nM); not active in vivo