Learn More
Cayman Chemical p-Nitrophenyl β-D-Galactopyranoside, Purity: ≥98%, CAS: 3150-24-1, 10 g
SDP
Catalog No. 502911927
Encompass_Preferred
A β-D-galactosidase chromogenic substrate; cleavage by β-D-galactosidase releases p-nitrophenol, which can be quantified by colorimetric detection at 420 nm as a measure of β-D-galactosidase activity
Catalog No. 50-291-1927 Supplier Cayman Chemical Supplier No. 3616010 G
A β-D-galactosidase chromogenic substrate; cleavage by β-D-galactosidase releases p-nitrophenol, which can be quantified by colorimetric detection at 420 nm as a measure of β-D-galactosidase activity