Learn More
Cayman Chemical p-Nitrophenyl β-D-Galactopyranoside, Purity: ≥98%, CAS: 3150-24-1, 5 g
SDP
Catalog No. 502911826
Encompass_Preferred
A β-D-galactosidase chromogenic substrate; cleavage by β-D-galactosidase releases p-nitrophenol, which can be quantified by colorimetric detection at 420 nm as a measure of β-D-galactosidase activity
Catalog No. 50-291-1826 Supplier Cayman Chemical Supplier No. 361605 G
A β-D-galactosidase chromogenic substrate; cleavage by β-D-galactosidase releases p-nitrophenol, which can be quantified by colorimetric detection at 420 nm as a measure of β-D-galactosidase activity