Learn More
Aviva Systems Biology Corp Pnkd Antibody - N-terminal region (ARP44997_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44997_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pnkd. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2210013N15Rik, 2810403H05Rik, AI854243, Brp17, MNCb-5687, MR-1, Tahccp2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-175