Learn More
Aviva Systems Biology Corp PNKP Antibody - N-terminal region (ARP52311_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52311_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PNKP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PNKP. Alternate names: PNK, MCSZ, EIEE10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-700