Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PNKP Human His-SUM 20ug
SDP

Catalog No. 5000195602
Encompass_Preferred

Recombinant human PNKP protein with N-terminal His and SUMO tags, expressed in E. coli and supplied as a lyophilized powder (20 μg). PNKP is a dual-activity polynucleotide kinase/phosphatase involved in DNA damage repair pathways (NHEJ and BER), removing 3'-phosphate groups and phosphorylating 5'-hydroxyl ends to prepare DNA for extension and ligation. Purity is >90% by reducing SDS-PAGE; formulation contains Tris-HCl, EDTA, and trehalose; store lyophilized at -20°C.

  • Recombinant human protein expressed in E. coli.
  • N-terminal His and SUMO tags for purification and stability.
  • Available as 20 μg lyophilized aliquot.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Lyophilized from 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0.
  • Store lyophilized at -20°C; reconstitute to ≥100 μg/mL recommended.

Catalog No. 50-001-95602 Supplier Medchemexpress LLC Supplier No. HYP7162020UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human PNKP protein with N-terminal His and SUMO tags, expressed in E. coli and supplied as a lyophilized powder (20 μg). PNKP is a dual-activity polynucleotide kinase/phosphatase involved in DNA damage repair pathways (NHEJ and BER), removing 3'-phosphate groups and phosphorylating 5'-hydroxyl ends to prepare DNA for extension and ligation. Purity is >90% by reducing SDS-PAGE; formulation contains Tris-HCl, EDTA, and trehalose; store lyophilized at -20°C.

  • Recombinant human protein expressed in E. coli.
  • N-terminal His and SUMO tags for purification and stability.
  • Available as 20 μg lyophilized aliquot.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Lyophilized from 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0.
  • Store lyophilized at -20°C; reconstitute to ≥100 μg/mL recommended.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.