Learn More
Medchemexpress LLC PNKP Human His-SUM 50ug
SDP
Catalog No. 5000196781
Encompass_Preferred
Recombinant human polynucleotide kinase/phosphatase (PNKP) is supplied lyophilized with N-terminal His and SUMO tags. Expressed in E. coli and provided at 50 μg with >90% purity, it is intended for biochemical assays, structural studies, and antibody validation and reflects the enzyme's roles in DNA repair (NHEJ and BER).
  • Lyophilized recombinant protein with N-terminal His and SUMO tags.
  • Expressed in E. coli and provided as a 50 μg vial for small-scale studies.
  • Purity greater than 90% by reducing SDS-PAGE for reproducible results.
  • Formulated with trehalose and EDTA to support stability during storage.
  • Suitable for enzymatic assays, structural analysis, and antibody validation.
  • Recommend aliquoting and storage at -20°C or -80°C for extended stability.
Catalog No. 50-001-96781 Supplier Medchemexpress LLC Supplier No. HYP7162050UG
Recombinant human polynucleotide kinase/phosphatase (PNKP) is supplied lyophilized with N-terminal His and SUMO tags. Expressed in E. coli and provided at 50 μg with >90% purity, it is intended for biochemical assays, structural studies, and antibody validation and reflects the enzyme's roles in DNA repair (NHEJ and BER).
  • Lyophilized recombinant protein with N-terminal His and SUMO tags.
  • Expressed in E. coli and provided as a 50 μg vial for small-scale studies.
  • Purity greater than 90% by reducing SDS-PAGE for reproducible results.
  • Formulated with trehalose and EDTA to support stability during storage.
  • Suitable for enzymatic assays, structural analysis, and antibody validation.
  • Recommend aliquoting and storage at -20°C or -80°C for extended stability.