Learn More
Aviva Systems Biology Corp PNLIP Antibody - C-terminal region (ARP33852_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33852_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PNLIP. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human PNLIP. Alternate names: PL, PTL, PNLIPDApplicationIHC, WB. Species reactivity: Human, Rabbit
Catalog No. 50-014-55173