Learn More
Aviva Systems Biology Corp PNLIP Antibody - C-terminal region (ARP33851_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33851_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PNLIP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PNLIP. Alternate names: PL, PTL, PNLIPDApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-55172