Learn More
Aviva Systems Biology Corp PNMA3 Antibody - middle region (ARP54946_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54946_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PNMA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PNMA3. Alternate names: MA3, MA5, MGC132756, MGC132758ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61378