Learn More
Aviva Systems Biology Corp PNO1 Antibody - N-terminal region (ARP62587_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62587_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PNO1. It was validated on Western Blot. . Alternate names: KHRBP1, RRP20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-720