Learn More
Cayman Chemical Panobinostat, Purity: ≥98%, CAS: 404950-80-7, 50 mg
SDP
Catalog No. 502954861
Encompass_Preferred
A pan-HDAC inhibitor (Kis = 0.6-31 nM for HDAC1-11); inhibits growth in a panel of 37 human or mouse lung cancer cell lines, including SCLC, NSCLC, and mesothelioma cells (IC50s = 4-175, 5-310, and 5-470 nM, respectively); induces tumor regression in BK-T and RG-1 SCLC mouse xenograft models at 20 mg/kg i.p.
Catalog No. 50-295-4861 Supplier Cayman Chemical Supplier No. 1328050 MG
A pan-HDAC inhibitor (Kis = 0.6-31 nM for HDAC1-11); inhibits growth in a panel of 37 human or mouse lung cancer cell lines, including SCLC, NSCLC, and mesothelioma cells (IC50s = 4-175, 5-310, and 5-470 nM, respectively); induces tumor regression in BK-T and RG-1 SCLC mouse xenograft models at 20 mg/kg i.p.