Learn More
Cayman Chemical PnobInostat-d4hydrochlorId 1mg
SDP
Catalog No. 502937803
Encompass_Preferred
An internal standard for the quantification of panobinostat by GC- or LC-MS
Catalog No. 50-293-7803 Supplier Cayman Chemical Supplier No. 285011 MG
An internal standard for the quantification of panobinostat by GC- or LC-MS