Learn More
Cayman Chemical Panobinostat-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), 5 mg
SDP
Catalog No. 502940626
Encompass_Preferred
An internal standard for the quantification of panobinostat by GC- or LC-MS
Catalog No. 50-294-0626 Supplier Cayman Chemical Supplier No. 285015 MG
An internal standard for the quantification of panobinostat by GC- or LC-MS