Learn More
Medchemexpress LLC pNP-ADPr disodium | 00-00-0 | 99.9% | C21H24N6Na2O16P2 | 10MG
SDP
Catalog No. 5000260384
Encompass_Preferred
pNP-ADPr disodium is a colorimetric substrate supplied as the disodium salt for enzymatic assays that measure poly(ADP-ribose) glycohydrolase (PARG) and ADP-ribosyl hydrolase 3 (ARH3) activity. It generates a chromogenic signal upon enzymatic cleavage, enabling continuous kinetic monitoring in biochemical experiments. The compound is provided as a solid for laboratory research use and is intended for assay development and enzyme characterization.
  • Colorimetric substrate for PARG and ARH3 assays.
  • Enables continuous kinetic measurement for real-time activity monitoring.
  • High purity for reproducible, low-background results.
  • Provided as the disodium salt to improve solubility in aqueous buffers.
  • Suitable for assay development and enzyme characterization experiments.
Catalog No. 50-002-60384 Supplier Medchemexpress LLC Supplier No. HY134354A10MG
pNP-ADPr disodium is a colorimetric substrate supplied as the disodium salt for enzymatic assays that measure poly(ADP-ribose) glycohydrolase (PARG) and ADP-ribosyl hydrolase 3 (ARH3) activity. It generates a chromogenic signal upon enzymatic cleavage, enabling continuous kinetic monitoring in biochemical experiments. The compound is provided as a solid for laboratory research use and is intended for assay development and enzyme characterization.
  • Colorimetric substrate for PARG and ARH3 assays.
  • Enables continuous kinetic measurement for real-time activity monitoring.
  • High purity for reproducible, low-background results.
  • Provided as the disodium salt to improve solubility in aqueous buffers.
  • Suitable for assay development and enzyme characterization experiments.