Learn More
Selleck Chemical LLC PNP-alpha-Glu E4014-1G
SDP
Catalog No. 5000041557
Encompass_Preferred
4-Nitrophenyl alpha-D-glucopyranoside is a chromogenic substrate in the -glucosidase inhibition assay
Catalog No. 50-000-41557 Supplier Selleck Chemical LLC Supplier No. E40141G
4-Nitrophenyl alpha-D-glucopyranoside is a chromogenic substrate in the -glucosidase inhibition assay