Learn More
Aviva Systems Biology Corp PNPLA2 Antibody - middle region (ARP58938_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58938_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PNPLA2. It was validated on Western Blot. . Alternate names: 1110001C14Rik, ATGL, DESNUTRIN, DKFZp667M109, FP17548, PEDF-R, TTS-2.2, TTS2ApplicationWB, IHC. Species reactivity: Human
Catalog No. 50-286-534