Learn More
Aviva Systems Biology Corp PNPLA3 Antibody - C-terminal region (ARP48936_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48936_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PNPLA3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human PNPLA3. Alternate names: ADPN, C22orf20, iPLA(2)epsilonApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-53567