Learn More
OriGene PNPLA3 Goat Polyclonal Antibody, 100 ug
SDP
Supplier:  OriGene TA303343
Encompass_Preferred
The protein encoded by this gene is a triacylglycerol lipase that mediates triacylglycerol hydrolysis in adipocytes. The encoded protein, which appears to be membrane bound, may be involved in the balance of energy usage/storage in adipocytes.
Catalog No. NC1737753