Learn More
Aviva Systems Biology Corp PNPLA4 Antibody - C-terminal region (ARP48639_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48639_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PNPLA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PNPLA4. Alternate names: DXS1283E, GS2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-59877