Learn More
Aviva Systems Biology Corp PNPLA5 Antibody - C-terminal region (ARP49011_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49011_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PNPLA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PNPLA5. Alternate names: 4833426H19Rik, GS2L, dJ388M5, dJ388M5.4ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59993